2-Amino-6-fluorobenzonitrile, Sigma-Aldrich®
Supplier: Merck
Synonyms:
2-Cyano-3-fluoroaniline
M-Clarity™ quality level = MQ100
429856-10G
429856-1G
SIAL429856-10GEA
179
EUR
SIAL429856-10G
SIAL429856-1G
2-Amino-6-fluorobenzonitrile, Sigma-Aldrich®
2-Amino-6-fluorobenzonitrile
Formula:
C₇H₅FN₂ MW: 136.13 g/mol Boiling Pt: 272 °C (1013 hPa) Melting Pt: 127 °C |
MDL Number:
MFCD00015477 CAS Number: 77326-36-4 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...