2-Amino-6-chlorobenzonitrile, Sigma-Aldrich®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ100

642843-1G 642843-5G
SIAL642843-1GEA 40.9 EUR
SIAL642843-1G SIAL642843-5G
2-Amino-6-chlorobenzonitrile, Sigma-Aldrich®
2-Amino-6-chlorobenzonitrile
Formula: C₇H₅ClN₂
MW: 152.58 g/mol
Boiling Pt: 301 °C (1013 hPa)
Melting Pt: 136…138 °C
MDL Number: MFCD00015546
CAS Number: 6575-11-7
EINECS: 229-501-4
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR