2,4-Difluorobenzonitrile, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
264326-25G 264326-5G
SIAL264326-25GEA 49.3 EUR
SIAL264326-25G SIAL264326-5G
2,4-Difluorobenzonitrile, Sigma-Aldrich®
2,4-Difluorobenzonitrile
Formula: C₇H₃F₂N
MW: 139.1 g/mol
Boiling Pt: 80…83 °C (22.5 mmHg)
Melting Pt: 46…47 °C
MDL Number: MFCD00009826
CAS Number: 3939-09-1
EINECS: 223-523-8
UN: 3439
ADR: 6.1,III