2-Fluoro-6-methoxybenzonitrile
Supplier: Thermo Scientific
TL00180.1
TL00180.10
TL00180.50
MAYBTL00180.1EA
0
EUR
MAYBTL00180.1
MAYBTL00180.10
MAYBTL00180.50
2-Fluoro-6-methoxybenzonitrile
2-Fluoro-6-methoxybenzonitrile
Formula:
C₈H₆FNO MW: 151.14 g/mol Boiling Pt: 245 °C (1013 hPa) Melting Pt: 93…95 °C |
MDL Number:
MFCD00042291 CAS Number: 94088-46-7 EINECS: 302-047-5 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...