DL-Alanine, Sigma-Aldrich®
Supplier: Merck
Synonyms: (±)-Alanine, DL-α-Alanine, H-DL-Ala-OH, α-Alanine
M-Clarity™ quality level = MQ300
W381810-10KG-K W381810-1KG-K W381810-5KG-K
SIALW381810-10KG-KEA 702 EUR
SIALW381810-10KG-K SIALW381810-1KG-K SIALW381810-5KG-K
DL-Alanine, Sigma-Aldrich®
DL-Alanine
Formula: C₃H₇NO₂
MW: 89.09 g/mol
Boiling Pt: 189
Melting Pt: 79
Density: 1.42 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064408
CAS Number: 302-72-7
EINECS: 206-126-4
Merck Index: 13,00203