DL-Alanine, Sigma-Aldrich®

Supplier: Merck

Synonyms: (±)-Alanine, DL-α-Alanine, H-DL-Ala-OH, α-Alanine

M-Clarity™ quality level = MQ200

A7502-1KG A7502-5KG
SIALA7502-1KGEA 171 EUR
SIALA7502-1KG SIALA7502-5KG
DL-Alanine, Sigma-Aldrich®
DL-Alanine
Formula: C₃H₇NO₂
MW: 89.09 g/mol
Boiling Pt: 189 °C (1013 hPa)
Melting Pt: 79 °C
Density: 1.42 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064408
CAS Number: 302-72-7
EINECS: 206-126-4
Merck Index: 13,00203

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR