2-Bromoisobutyric acid, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
306851-100G
306851-25G
SIAL306851-100GEA
128
EUR
SIAL306851-100G
SIAL306851-25G
2-Bromoisobutyric acid, Sigma-Aldrich®
2-Bromoisobutyric acid
Formula:
C₄H₇BrO₂ Boiling Pt: 198…200 °C (1013 hPa) Melting Pt: 48 °C Density: 1.43 g/cm³ (20 °C) |
MDL Number:
MFCD00004168 CAS Number: 2052-01-9 EINECS: 218-139-2 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...