2-Chloro-5-(chloromethyl)pyridine 96%
Supplier: Thermo Scientific
Danger
L19284.06 L19284.14
L19284.06EA 46.6 EUR
L19284.06 L19284.14
2-Chloro-5-(chloromethyl)pyridine 96%
2-Chloro-5-(chloromethyl)pyridine
Purity: 96% CAS No.: 70258-18-3 Molecular Formula: C6H5Cl2N Formula Weight: 162.02 Flash Point: 110°(230°F) Melting Point: 37-42° Beilstein Reference: 1635690 UN No.: 3261 MDL No.: MFCD00125366
Formula: C₆H₅Cl₂N
MW: 162.02 g/mol
Melting Pt: 37…42 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00125366
CAS Number: 70258-18-3
UN: 3261
ADR: 8,II