β-Alanine, Sigma-Aldrich®
Supplier: Merck
Synonyms: H-β-Ala-OH, 3-Aminopropionic acid, H-Bala-OH, 3-Aminopropanoic acid
M-Clarity™ quality level = MQ300
05160-1KG 05160-250G 05160-50G
SIAL05160-1KGEA 163 EUR
SIAL05160-1KG SIAL05160-250G SIAL05160-50G
β-Alanine, Sigma-Aldrich®
β-Alanine
Formula: C₃H₇NO₂
MW: 89.09 g/mol
Boiling Pt: 201 °C (1013 hPa)
Melting Pt: 94 °C
Density: 1.44 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008200
CAS Number: 107-95-9
EINECS: 203-536-5
Merck Index: 13,00204