β-Alanine, Sigma-Aldrich®
Supplier: Merck
Synonyms:
H-β-Ala-OH, 3-Aminopropionic acid, H-Bala-OH, 3-Aminopropanoic acid
M-Clarity™ quality level = MQ200
A9920-100G
A9920-10MG
SIALA9920-100GEA
47.7
EUR
SIALA9920-100G
SIALA9920-10MG
β-Alanine, Sigma-Aldrich®
β-Alanine
Formula:
C₃H₇NO₂ MW: 89.09 g/mol Boiling Pt: 201 °C (1013 hPa) Melting Pt: 94 °C Density: 1.44 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008200 CAS Number: 107-95-9 EINECS: 203-536-5 Merck Index: 13,00204 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...