Cholesterol, Sigma-Aldrich®
Supplier: Merck
Synonyms: Cholest-5-en-3-ol, Cholesterin, (3Beta)-Cholest-5-en-3-ol
MQ200
C8667-500MG C8667-100G C8667-1G C8667-25G C8667-5G
SIALC8667-500MGEA 44.1 EUR
SIALC8667-500MG SIALC8667-100G SIALC8667-1G SIALC8667-25G SIALC8667-5G
Cholesterol, Sigma-Aldrich®
Cholesterol
Formula: C₂₇H₄₆O
MW: 386.66 g/mol
Boiling Pt: 360
Melting Pt: 147…150 °C
Density: 1.07 g/cm³ (20 °C)
Storage Temperature: Freezer
MDL Number: MFCD00003646
CAS Number: 57-88-5
EINECS: 200-353-2
Merck Index: 13,02221