(±)-3-Hydroxybutyric acid, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 3-Hydroxybutyric acid, DL-3-Hydroxybutyric Acid, (±)-3-Hydroxybutanoic acid
M-Clarity™ quality level = MQ100
166898-1G 166898-5G
SIAL166898-1GEA 73.9 EUR
SIAL166898-1G SIAL166898-5G
(±)-3-Hydroxybutyric acid, Sigma-Aldrich®
(±)-3-Hydroxybutyric acid
Formula: C₄H₈O₃
MW: 104.11 g/mol
Boiling Pt: 269 °C (1013 hPa)
Melting Pt: 33…34 °C
Density: 1.26 g/cm³ (20 °C)
MDL Number: MFCD00004546
CAS Number: 300-85-6
EINECS: 206-099-9
Merck Index: 12,04861