3-Pyridineboronic acid, Sigma-Aldrich®

Supplier: Merck

Synonyms: 3-Pyridinylboronic acid, Pyridin-3-ylboronic acid, Pyridine-3-boronic acid

M-Clarity™ quality level = MQ100

512125-1G 512125-25G 512125-5G
SIAL512125-1GEA 96.5 EUR
SIAL512125-1G SIAL512125-25G SIAL512125-5G
3-Pyridineboronic acid, Sigma-Aldrich®
3-Pyridineboronic acid
Formula: C₅H₆BNO₂
MW: 122.92 g/mol
MDL Number: MFCD00674177
CAS Number: 1692-25-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR