5α-Cholestane, Sigma-Aldrich®
Supplier: Merck
MQ200
C8003-1G
C8003-100MG
C8003-5G
SIALC8003-1GEA
290
EUR
SIALC8003-1G
SIALC8003-100MG
SIALC8003-5G
5α-Cholestane, Sigma-Aldrich®
5α-Cholestane
Formula:
C₂₇H₄₈ MW: 372.68 g/mol Boiling Pt: 441 Melting Pt: 79 Density: 0.9133 g/cm³ (80 °C) |
MDL Number:
MFCD00066412 CAS Number: 481-21-0 EINECS: 207-562-8 Merck Index: 12,02254 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...