Barium chloride 0.25 mol/l (0.5 N) in aqueous solution Reag. Ph. Eur. 1009301
Supplier: VWR Chemicals
87783.290EA 338 EUR
87783.290
Barium chloride 0.25 mol/l (0.5 N) in aqueous solution Reag. Ph. Eur. 1009301
Barium chloride
Informace pro objednávání: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.
Formula: BaCl₂
MW: 208,24 g/mol
Density: 1,034 g/cm³ (20 °C)
MDL Number: MFCD00003445
UN: 3287
ADR: 6.1,III