Barium hydroxide octahydrate 99.95%
Supplier: Thermo Scientific
Danger
377820050 377820250
ACRO377820050EA 52.2 EUR
ACRO377820050 ACRO377820250
Barium hydroxide octahydrate 99.95%
Barium hydroxide octahydrate
Formula: Ba(OH)₂·8H₂O
MW: 315.47 g/mol
Melting Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00149152
CAS Number: 12230-71-6
EINECS: 241-234-5
UN: 3262
ADR: 8,II
Order Now
Specification Test Results
Appearance White Crystalline powder or crystals or flakes and/or chunks
Total trace metal impurities ≤0.05 %