Immersion Oil, Type N
Supplier: NIKON
MXA22203 MXA22202
NIKOMXA22203EA 60.5 EUR
NIKOMXA22203 NIKOMXA22202
Immersion Oil, Type N
Stains Microscopy and staining reagents
Type N is a general purpose oil recommended for transmitted light applications, including brightfield, darkfield, DIC and phase contrast.
These oils are tested using Nikon objectives and therefore the performance cannot be guaranteed against lenses manufactured by other companies. The refractive indices of other manufactured oils may differ which will impair the results gained from using these with Nikon objectives. It is also important to note that oils should not be mixed as this will impair the performance.