Cesium hydroxide monohydrate, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
562505-1KG 562505-5KG
SIAL562505-1KGEA 548 EUR
SIAL562505-1KG SIAL562505-5KG
Cesium hydroxide monohydrate, Sigma-Aldrich®
Cesium hydroxide monohydrate
Formula: HCsO·1H₂O
MW: 167.93 g/mol
Melting Pt: 204.85 °C
Density: 3.51 g/cm³ (24.85 °C)
MDL Number: MFCD00149664
CAS Number: 35103-79-8
EINECS: 244-344-1
UN: 3262
ADR: 8,II
Merck Index: 12,02055