Cesium hydroxide hydrate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Cesium hydroxide, hydrate
C8518-10G C8518-250G C8518-50G
SIALC8518-10GEA 99.1 EUR
SIALC8518-10G SIALC8518-250G SIALC8518-50G
Cesium hydroxide hydrate, Sigma-Aldrich®
Cesium hydroxide hydrate
Formula: HCsO·H₂O
MW: 167.93 g/mol
MDL Number: MFCD00010964
CAS Number: 12260-45-6
EINECS: 244-344-1
UN: 2682
ADR: 8,II