Cesium hydroxide 50% in aqueous solution, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
232068-100G 232068-500G
SIAL232068-100GEA 124 EUR
SIAL232068-100G SIAL232068-500G
Cesium hydroxide 50% in aqueous solution, Sigma-Aldrich®
Cesium hydroxide
Formula: CsOH
MDL Number: MFCD00010964
CAS Number: 21351-79-1
EINECS: 244-344-1
UN: 2681
ADR: 8,II
Merck Index: 13,02022