Cesium hydroxide 50% in aqueous solution, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
232041-10G 232041-200G 232041-50G
SIAL232041-10GEA 51 EUR
SIAL232041-10G SIAL232041-200G SIAL232041-50G
Cesium hydroxide 50% in aqueous solution, Sigma-Aldrich®
Cesium hydroxide
Formula: CsOH
MDL Number: MFCD00010964
CAS Number: 21351-79-1
EINECS: 244-344-1
UN: 2681
ADR: 8,II
Merck Index: 13,02022