Chloramphenicol, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Chloromycetin®, Tega-Cetin, (R,R)-Chloramphenicol
M-Clarity™ quality level = MQ300
C0857-100G C0857-25G C0857-500G
SIALC0857-100GEA 414 EUR
SIALC0857-100G SIALC0857-25G SIALC0857-500G
Chloramphenicol, Sigma-Aldrich®
Chloramphenicol
Formula: Cl₂CHCONHCH(CH₂OH)CH(OH)C₆H₄NO₂
MW: 323.13 g/mol
Boiling Pt: 645 °C (1013 hPa)
Melting Pt: 150 °C
Density: 1.49 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00078159
CAS Number: 56-75-7
EINECS: 200-287-4
Merck Index: 13,02087