Chloramphenicol, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
Chloromycetin®, Tega-Cetin, (R,R)-Chloramphenicol
M-Clarity™ quality level = MQ200
C1919-100G
C1919-25G
C1919-5G
SIALC1919-100GEA
596
EUR
SIALC1919-100G
SIALC1919-25G
SIALC1919-5G
Chloramphenicol, Sigma-Aldrich®
Chloramphenicol
Formula:
Cl₂CHCONHCH(CH₂OH)CH(OH)C₆H₄NO₂ MW: 323.13 g/mol Boiling Pt: 645 °C (1013 hPa) Melting Pt: 150 °C Density: 1.49 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00078159 CAS Number: 56-75-7 EINECS: 200-287-4 Merck Index: 13,02087 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...