Thiram, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
Tetramethylthiuram disulphide
M-Clarity™ quality level = MQ100
T24201-100G
T24201-5G
SIALT24201-100GEA
88.4
EUR
SIALT24201-100G
SIALT24201-5G
Thiram, Sigma-Aldrich®
Thiram
Formula:
C₆H₁₂N₂S₄ MW: 240.44 g/mol Boiling Pt: 307 Melting Pt: 148…152 °C Density: 1.426 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00008325 CAS Number: 137-26-8 EINECS: 205-286-2 UN: 2811 ADR: 6.1 II Merck Index: 12,09510 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...