MyLab™ Electrophoresis Sampler Kit
Supplier: Edvotek
MyLab™
EDVO100EA 94.1 EUR
EDVO100
MyLab™ Electrophoresis Sampler Kit
Electrophoresis Systems Horizontal Gel Electrophoresis Systems
In this experiment kit, students will explore four unique experiments that utilise agarose gel electrophoresis to separate DNA samples. The DNA banding patterns will be analysed within scenarios that examine forensic, medical, and scientific investigations.
These kits can be stored at room temperature. Storage of Ready-to-Load QuickStrip™ samples in the refrigerator is recommended.
Required: DNA electrophoresis, micropipettes: 5 to 50 µl (Optional), white light box, blue light transilluminator (Optional) and microwave or hot plate.
Time required: Complete in 45 minutes
Delivery information: Kit includes: Instructions, Ready-to-Load QuickStrip™ DNA samples, UltraSpec-Agarose™, electrophoresis buffer (50X), practice gel loading solution, FlashBlue™ DNA stain and SYBR™ safe stain.