Clofilium tosylate ≥97% (by HPLC), white solid
Supplier: ENZO LIFE SCIENCES
Warning
Synonyms: 4-Chloro-N,N-diethyl-N-heptylbenzenebutanaminium . tosylate
ALX550242M100 ALX550242M250
ENZOALX550242M100EA 1180 EUR
ENZOALX550242M100 ENZOALX550242M250
Clofilium tosylate ≥97% (by HPLC), white solid
Clofilium tosylate
K⁺ channel blocker. Antiarrhytmic. Cardiac depressant.
Formula: C₂₈H₄₄ClNO₃S
Storage Temperature: Ambient
CAS Number: 92953-10-1
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Purity (HPLC) ≥97%
Identity Determined by 1H-NMR