Oakton® Conductivity and TDS Standards
Supplier: Avantor
Oakton®
VWRU76338-556EA 0 EUR
VWRU76338-556
Oakton® Conductivity and TDS Standards
Standards Conductivity Standards
Ideal for laboratory and field analysis.
  • Traceable to NIST standard reference materials
  • Solutions help you calibrate conductivity instruments to ensure accuracy
  • Standard values given at 25 °C
Oakton conducitivity and TDS standards feature temperature corrections, KCl, NaCL, and 442 conversions printed directly on the bottle for ease of use (excluded on 60-mL bottle, but included in insert).
Certifikace: These standards are manufactured under ISO 9000 quality guidelines and traceable to NIST standards.
Upozornení: If unopened, shelf life is one year. If opened, shelf life if 4 months.