5-Carboxyfluorescein ≥98% (by HPLC), Ultrapure

Supplier: ENZO LIFE SCIENCES AG

ENZOENZ52051EA 134.2 EUR
ENZOENZ52051
5-Carboxyfluorescein ≥98% (by HPLC), Ultrapure
5-Carboxyfluorescein

Amine reactive dye


5-FAM is the purified single isomer of 5,6-carboxyfluorescein and is used for labeling peptides, proteins and nucleotides through the interaction of carboxylic acid with primary amines. It has been principally used to develop a variety of green fluorescent reagents and small fluorescent molecules due to its relatively high absorbance characteristics, excellent fluorescence quantum yield, and good water solubility. The probe has also found application in studies of liposome-cell, cell-cell, and liposome-liposome interactions and in related studies on lipid bilayer structures. Wavelength Maxima: Excitation 492 nm, Emission 518 nm.

Formula: C₂₁H₁₂O₇.xH₂O
MW: 376,32 g/mol (anhy)
Storage Temperature: Freezer
MDL Number: MFCD00036874
CAS Number: 76823-03-5
https://si.vwr.com/stibo/web/std.lang.all/15/92/45401592.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR