N-Palmitoyl ceramide ≥98% (by TLC)
Supplier: ENZO LIFE SCIENCES
Synonyms: D-erythro-Sphingosine, N-Palmitoyl-, N-Palmitoyl-D-erythro-sphingosine, C16 Ceramide, N-Hexadecanoyl-D-erythro-sphingosine
BMLSL1150005 BMLSL1150025
ENZOBMLSL1150005EA 84.7 EUR
ENZOBMLSL1150005 ENZOBMLSL1150025
N-Palmitoyl ceramide ≥98% (by TLC)
N-Palmitoyl ceramide
Endogenous ceramide.
An abundant molecular species of endogenous ceramide. It induces phosphorylation of epidermal growth factor receptor on Thr⁶⁶⁹ in A-431 cells by stimulation of ceramide-activated protein kinase and activates NF-κB at concentrations as low as 50 nM. Activates PKCζ.
Soluble in 100% ethanol (5 mg/ml warm).
Formula: C₃₄H₆₇NO₃
MW: 537.914 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00871748
CAS Number: 24696-26-2
Order Now
Specification Test Results
Purity ≥98% (TLC)