C8 ceramide ≥98% (by TLC)
Supplier: ENZO LIFE SCIENCES
BMLSL1120005 BMLSL1120025
ENZOBMLSL1120005EA 66 EUR
ENZOBMLSL1120005 ENZOBMLSL1120025
C8 ceramide ≥98% (by TLC)
C8 ceramide
Ceramide-activated protein kinase activator.
A biologically active, cell-permeable, but nonphysiologic ceramide analog. It induces phosphorylation of epidermal growth factor receptor on Thr-669 in A-431 cells by stimulation of ceramide-activated protein kinase and induces apoptosis.
Soluble in DMSO (25 mg/ml) or 100% ethanol (25 mg/ml).
Formula: C₂₆H₅₁NO₃
MW: 425.698 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00236496
CAS Number: 74713-59-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=777f30a2b1ae3a32b5a2ec1027aebdb691050282&vid=9cdc126e319aa4dbe640d22b0c8c678f7a6d6fd9&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity ≥98% (TLC)