N-[2-(Cyclohexyloxy)-4-nitrophenyl]methanesulphonamide ≥98% (by HPLC)
Supplier: ENZO LIFE SCIENCES
Synonyms: N-(2-(Cyclohexyloxy)-4-nitrophenyl)methanesulphonamide, NS-398, N-[2-(Cyclohexyloxy)-4-nitrophenyl]methanesulphonamide
BMLEI2610005 BMLEI2610025
ENZOBMLEI2610005EA 114 EUR
ENZOBMLEI2610005 ENZOBMLEI2610025
N-[2-(Cyclohexyloxy)-4-nitrophenyl]methanesulphonamide ≥98% (by HPLC)
N-[2-(Cyclohexyloxy)-4-nitrophenyl]methanesulphonamide
Cyclooxygenase inhibitor. Selectively inhibits PGH synthase-2 (IC50=1 µM) with no inhibition of PGH synthase-1 at concentrations up to 100 µM. Protects neuronal cultures from LPS-induced neurotoxicity.
  • COX inhibitor
Cyclooxygenase inhibitor. Selectively inhibits PGH synthase-2 (IC50=1 µM) with no inhibition of PGH synthase-1 at concentrations up to 100 µM. Protects neuronal cultures from LPS-induced neurotoxicity.
Soluble in 100% ethanol (warm; sonication), DMSO (25mg/ml) or dimethyl formamide. Insoluble in water.
Formula: C₁₃H₁₈N₂O₅S
MW: 314.356 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00882995
CAS Number: 123653-11-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f9213d5b3ded2e62b16d0d1929536f2b404d5630&vid=d2c7ad5aad8b3c6dc3d3d57bfb72e86684ee90e3&prid=web&clid=SAPDAM