8-Br-cGMP 96 HPLC_ASSAY_METHOD
Supplier: ENZO LIFE SCIENCES
BMLCN2050010 BMLCN2050050
ENZOBMLCN2050010EA 86.9 EUR
ENZOBMLCN2050010 ENZOBMLCN2050050
8-Br-cGMP 96 HPLC_ASSAY_METHOD
8-Br-cGMP
A cell-permeable cGMP analog which is more resistant to phosphodiesterases than cGMP and which preferentially activates cGMP-dependent protein kinase. It inhibits thrombin stimulated arachidonic acid release in human platelets.
  • PKG activator
White to off-white powder
Soluble in water (25mg/ml) or aqueous buffers. Sparingly soluble in methanol (4mM). Insoluble in DMSO, dimethyl formamide and 96% ethanol.
Formula: C₁₀H₁₂BrN₅NaO₈P
MW: 464.101 g/mol
Storage Temperature: Freezer
CAS Number: 51116-01-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6ae97b73b1204d05a6117abe87242bcd80ec00d8&vid=3f6827122ac1cb58dff9e2e731bec69ef589ce9f&prid=web&clid=SAPDAM