25-Hydroxyvitamin D3 98 HPLC_ASSAY_METHOD, solid_white to off white
Supplier: ENZO LIFE SCIENCES
Danger
Synonyms: Calcifediol, (R,Z)-3-(2-((1R,3aS,7aR,E)-1-((R)-6-hydroxy-6-methylheptan-2-yl)-7a-methyloctahydro-4H-inden-4-ylidene)ethylidene)-4-methylenecyclohexan-1-ol
BMLDM1000001 BMLDM1000005
ENZOBMLDM1000001EA 71.5 EUR
ENZOBMLDM1000001 ENZOBMLDM1000005
25-Hydroxyvitamin D3 98 HPLC_ASSAY_METHOD, solid_white to off white
25-Hydroxyvitamin D3
Precursor to 1α,25-dihydroxyvitamin D₃
Intermediate in the biosynthesis of 1alpha,25-dihydroxyvitamin D3. Major circulating metabolite of vitamin D3.
Soluble in 100% ethanol.
Formula: C₂₇H₄₄O₂
MW: 400.6 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00867077
CAS Number: 19356-17-3
UN: 2811
ADR: 6.1 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e3ec23887b3d8e238fd5ef1c6f5940df673eb623&vid=ce4a390b2c87e0097d88cb0406310e7e5e60a035&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Identity Determined by IR
Purity (HPLC) ≥98%