7-HC-arachidonate 98 HPLC_ASSAY_METHOD
Supplier: ENZO LIFE SCIENCES
Warning
BMLST5020010 BMLST5020050
ENZOBMLST5020010EA 110 EUR
ENZOBMLST5020010 ENZOBMLST5020050
7-HC-arachidonate 98 HPLC_ASSAY_METHOD
7-HC-arachidonate
Phospholipase substrate.
PLA₂ fluorogenic substrate for use in assays with phospholipid and Ca²⁺. For use with purified enzyme, as this substrate is nonspecific. Ideal for drug discovery, HTS, and similar applications. Ex.: 360 nm, Em.: 460 nm
Soluble in DMSO (50 mg/ml) or hexane. Do not use ethanol as it may degrade product.
Formula: C₂₉H₃₆O₄
MW: 448.6 g/mol
Storage Temperature: Freezer
CAS Number: 161180-11-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4784c05fee73d024c84c1bf125257a8a8e1ebce4&vid=acc2312f02fbdcac130d528cb8ae6a19e511bc60&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity ≥98% (HPLC)