RO 20-1724 99 TLC_ASSAY_METHOD
Supplier: ENZO LIFE SCIENCES
BMLEI1170100 BMLEI1171000
ENZOBMLEI1170100EA 147 EUR
ENZOBMLEI1170100 ENZOBMLEI1171000
RO 20-1724 99 TLC_ASSAY_METHOD
RO 20-1724
Selective, potent cAMP-specific phosphodiesterase inhibitor (PDE IV, IC50=2µM). Inhibits superoxide generation and platelet aggregation stimulated by arachidonic acid. Also inhibits fMLP-induced neutrophil adhesion to vascular endothelial cells.
  • PDE inhibitor
White to off-white solid.
Soluble in 100% ethanol (40mg/ml) or DMSO (200 mg/ml); insoluble in water.
Formula: C₁₅H₂₂N₂O₃
MW: 278.3 g/mol
Storage Temperature: Ambient
CAS Number: 29925-17-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eda329397442e9c5b37101d7fea8a78a8efc1972&vid=80057058e4443d4615d3f7032db610fbe38c3fd0&prid=web&clid=SAPDAM