NSC 23766 ≥99% (by HPLC)
Supplier: ENZO LIFE SCIENCES
ENZCHM1160005 ENZCHM1160010
ENZOENZCHM1160005EA 121 EUR
ENZOENZCHM1160005 ENZOENZCHM1160010
NSC 23766 ≥99% (by HPLC)
NSC 23766
Cell-permeable, reversible, and selective Rac1 inhibitor.
NSC 23766 has been used to investigate the role of Rac1 in such diverse cellular functions as stem cell mobilization, epithelial cell migration, angiogenesis, leukemia cell migration and growth.
Formula: C₂₄H₃₅N₇·3HCl
MW: 531 g/mol
MDL Number: MFCD08703096
CAS Number: 733767-34-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=83e59d4efe7d4e39249ff7b8ee22027a2a79014d&vid=f0fc596b417f30abff31d26df1555ae9c2ab86ac&prid=web&clid=SAPDAM