Dimethyl-bisphenol A ≥98% (by TLC)
Supplier: ENZO LIFE SCIENCES
Synonyms: 4,4'-(Propane-2,2-diyl)bis(methoxybenzene)
BMLGR3390010 BMLGR3390050
ENZOBMLGR3390010EA 114 EUR
ENZOBMLGR3390010 ENZOBMLGR3390050
Dimethyl-bisphenol A ≥98% (by TLC)
Dimethyl-bisphenol A
A HIF-1α degradation promotor.
  • HIF-1α inhibitor
White solid.
Soluble in DMSO (>25 mg/ml) or ethanol (>25 mg/ml)
Formula: C₁₇H₂₀O₂
MW: 256.3 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00984727
CAS Number: 1568-83-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a4fe6ed400bc1ccbbfccd206127ad89c3b365c0&vid=53366f7879f0f7c1b29f3caa97a35fbd5558eae9&prid=web&clid=SAPDAM