L(+)-Arginine

Supplier: ENZO LIFE SCIENCES AG
Warning

ENZOALX101004G025EA 50.6 EUR
ENZOALX101004G025
L(+)-Arginine
L(+)-Arginine
Formula: C₆H₁₄N₄O₂
MW: 174,2 g/mol
Boiling Pt: 368 °C (1013 hPa)
Melting Pt: 238 °C
Density: 1,3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002635
EINECS: 200-811-1
Merck Index: 13,00785

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR