T0901317 ≥98% (by TLC)
Supplier: ENZO LIFE SCIENCES
Synonyms: N-(4-(1,1,1,3,3,3-Hexafluoro-2-hydroxypropan-2-yl)phenyl)-N-(2,2,2-trifluoroethyl)benzenesulphonamide
ALX270309M010 ALX270309M050
ENZOALX270309M010EA 56.1 EUR
ENZOALX270309M010 ENZOALX270309M050
T0901317 ≥98% (by TLC)
T0901317
LXR activator
Potent agonist (EC50=50nM) of the orphan nuclear hormone receptor (liver X receptor; LXR). Inhibitor of human sex hormone-dependent cancer cell lines.
Formula: C₁₇H₁₂F₉NO₃S
MDL Number: MFCD03412028
CAS Number: 293754-55-9