CVT-313 ≥98% (by HPLC)
Supplier: ENZO LIFE SCIENCES
Synonyms: 2,2'-((9-isopropyl-6-((4-methoxybenzyl)amino)-9H-purin-2-yl)azanediyl)diethanol
ALX270393M001 ALX270393M005
ENZOALX270393M001EA 120 EUR
ENZOALX270393M001 ENZOALX270393M005
CVT-313 ≥98% (by HPLC)
CVT-313
CDK/cyclin inhibitor
Specific inhibitor of CDK2/cyclin A (IC50=500nM) and CDK2/cyclin E (IC50=500nM). Also inhibits CDK1/cyclin B (IC50=4.2µM) and CDK4/cyclin D1 (IC50=215µM).
Formula: C₂₀H₂₈N₆O₃
MDL Number: MFCD06411412
CAS Number: 199986-75-9