Polymyxin B sulphate
Supplier: ENZO LIFE SCIENCES
Warning
ALX380040G001 ALX380040G005
ENZOALX380040G001EA 120 EUR
ENZOALX380040G001 ENZOALX380040G005
Polymyxin B sulphate
Polymyxin B sulphate
PKC inhibitor
Antibiotic. Inhibitor of protein kinase C (PKC). Breaks the bacterial membrane by incorporating into the phospholipid of the outer membrane and activating phospholipase.
Isolated from Bacillus polymyxa.
Formula: C₅₅H₉₆N₁₆O₁₃·2H₂SO₄
MW: 1385.61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00131991
CAS Number: 1405-20-5
EINECS: 215-774-7