3,3',4',5,7-Pentahydroxyflavylium chloride ≥97% (by HPLC)
Supplier: ENZO LIFE SCIENCES
Warning
Synonyms: 2-(3,4-Dihydroxyphenyl)-3,5,7-trihydroxychromenylium chloride, Cyanidin chloride
ALX385003M010 ALX385003M050
ENZOALX385003M010EA 372 EUR
ENZOALX385003M010 ENZOALX385003M050
3,3',4',5,7-Pentahydroxyflavylium chloride ≥97% (by HPLC)
3,3',4',5,7-Pentahydroxyflavylium chloride
Nitric oxide (NO) scavenger.
Antioxidant flavonoid.
Soluble in methanol or 0.1% hydrochloric acid.
Formula: C₁₅H₁₁O₆Cl
MW: 322.7 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00017582
CAS Number: 528-58-5
EINECS: 208-438-6
Merck Index: 14,02687
Order Now
Specification Test Results
Purity ≥97% (HPLC)
Identity Determined by ¹H-NMR, ¹³C-NMR and MS