Guanosine-5'-O-(3-thiotriphosphate) tetralithium salt ≥85% (by HPLC)
Supplier: ENZO LIFE SCIENCES
Warning
ALX480056M001 ALX480056M005
ENZOALX480056M001EA 85.8 EUR
ENZOALX480056M001 ENZOALX480056M005
Guanosine-5'-O-(3-thiotriphosphate) tetralithium salt ≥85% (by HPLC)
Guanosine-5'-O-(3-thiotriphosphate) tetralithium salt
Nonhydrolysable GTP analog.
A nonhydrolysable analog of guanosine-5’-O-triphosphate (GTP). Stimulates inositol trisphosphate formation in membranes.
Soluble in water (25 mg/ml).
Formula: C₁₀H₁₂N₅O₁₃P₃S·4Li
MW: 563 g/mol
Storage Temperature: Freezer
CAS Number: 94825-44-2
Order Now
Specification Test Results
Purity (HPLC) ≥85%