Kolhicin, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

SIALC9754-1GEA 114 EUR
SIALC9754-1G SIALC9754-5G SIALC9754-500MG SIALC9754-100MG
Kolhicin, Sigma-Aldrich®
Kolhicin
Formula: C₂₂H₂₅NO₆
MW: 399,44 g/mol
Melting Pt: 154…156 °C
Density: 1,32 g/cm³ (20 °C)
MDL Number: MFCD00078484
UN: 1544
ADR: 6.1,I
Merck Index: 12,02536

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR