4-Bromo-2,6-di-tert-butylphenol, Sigma-Aldrich®
Supplier: Merck
Synonyms:
1-Bromo-3,5-di(dimethylethyl)-4-hydroxybenzene
M-Clarity™ quality level = MQ100
394378-100G
394378-25G
SIAL394378-100GEA
276
EUR
SIAL394378-100G
SIAL394378-25G
4-Bromo-2,6-di-tert-butylphenol, Sigma-Aldrich®
4-Bromo-2,6-di-tert-butylphenol
Formula:
C₁₄H₂₁BrO MW: 285.22 g/mol Boiling Pt: 104…107 °C (1 mmHg) Melting Pt: 83 °C |
MDL Number:
MFCD00051598 CAS Number: 1139-52-2 EINECS: 214-521-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...