2-Methoxystyrene, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

563064-1G 563064-5G
SIAL563064-1GEA 60.8 EUR
SIAL563064-1G SIAL563064-5G
2-Methoxystyrene, Sigma-Aldrich®
2-Methoxystyrene
Formula: C₉H₁₀O
MW: 134.18 g/mol
Boiling Pt: 197 °C (1013 hPa)
Melting Pt: 29 °C
Density: 0.999 g/cm³ (20 °C)
CAS Number: 612-15-7
EINECS: 210-294-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR