4-Methylstyrene, Sigma-Aldrich®

Supplier: Merck

Synonyms: p-Vinyltoluene, p-Methylstyrene, 4-Vinyltoluene

M-Clarity™ quality level = MQ100

M80806-100ML M80806-10ML M80806-500ML
SIALM80806-100MLEA 56.8 EUR
SIALM80806-100ML SIALM80806-10ML SIALM80806-500ML
4-Methylstyrene, Sigma-Aldrich®
4-Methylstyrene
Formula: C₉H₁₀
MW: 118.18 g/mol
Boiling Pt: 173 °C (1013 hPa)
Melting Pt: –34 °C
Density: 0.9383 g/cm³ (0 °C)
MDL Number: MFCD00008621
CAS Number: 622-97-9
EINECS: 210-762-8
UN: 2618
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR