Methyl red, Sigma-Aldrich®
Supplier: Merck
Synonyms:
2-(4-Dimethylaminophenylazo)benzoic acid, 2-((4-(Dimethylamino)phenyl)diazenyl)benzoic acid
M-Clarity™ quality level = MQ200
250198-100G
250198-25G
SIAL250198-100GEA
214
EUR
SIAL250198-100G
SIAL250198-25G
Methyl red, Sigma-Aldrich®
Methyl red
Formula:
C₁₅H₁₅N₃O₂ Melting Pt: 178…182 °C Density: 1.31 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002425 CAS Number: 493-52-7 EINECS: 207-776-1 Merck Index: 12,06199 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...