2-Acetyl-4-methylthiophene, Sigma-Aldrich®
Supplier: Merck
Synonyms:
1-(4-Methyl-2-thienyl)-1-ethanone, 1-(4-Methylthiophen-2-yl)ethanone
M-Clarity™ quality level = MQ100
633046-25G
633046-5G
SIAL633046-25GEA
336
EUR
SIAL633046-25G
SIAL633046-5G
2-Acetyl-4-methylthiophene, Sigma-Aldrich®
2-Acetyl-4-methylthiophene
Formula:
C₇H₈OS MW: 140.21 g/mol Boiling Pt: 108…109 °C (14 mmHg) |
MDL Number:
MFCD00027395 CAS Number: 13679-73-7 EINECS: 237-180-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...