Bromocresol green sodium salt 0.04% (w/v) in aqueous solution
Supplier: Thermo Scientific
Synonyms: Bromocresol green water soluble, Sodium 2,6-dibromo-4-(3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-2,2-dioxido-3H-benzo[d][1,2]oxathiol-3-yl)-3-methylphenolate
38696.AE 38696.AP
38696.AEEA 39.6 EUR
38696.AE 38696.AP
Bromocresol green sodium salt 0.04% (w/v) in aqueous solution
Bromocresol green sodium salt
CAS No.: 62625-32-5 Molecular Formula: C21H13Br4NaO5S Formula Weight: 720.01 Physical Form: Liquid Appearance: Green
Formula: C₂₁H₁₃Br₄NaO₅S
MW: 720 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00148898
CAS Number: 62625-32-5
EINECS: 263-657-4